missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one, Mikromol
Specifications
Specifications
| Chemical Name or Material | 3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one |
| CAS | 82543-15-5 |
| Analyte or Component Names | 3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H28 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3'H-Cycloprop[15,16]androsta-6,15-dien-17-one, 15,16-dihydro-3,5-dihydroxy-, (3β,5β,15α,16α)-, 3β,5-Dihydroxy-15β,16β-methylene-5β-androst-6-en-17-one |
| InChI Formula | InChI=1S/C20H28O3/c1-18-6-5-15-12(16(18)13-9-14(13)17(18)22)4-8-20(23)10-11(21)3-7-19(15,20)2/h4,8,11-16,21,23H,3,5-7,9-10H2,1-2H3/t11-,12+,13+,14-,15-,16+,18-,19+,20-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](C=C[C@]4(O)C[C@@H](O)CC[C@]34C)[C@@H]1[C@@H]5C[C@@H]5C2=O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction