Learn More
Nalgene 32 oz. Narrow Mouth Bottle
Description
- Specially designed lip ideal for keeping the flow, without the stress of a mess
- Tough and guaranteed for life
Specifications
Specifications
| Capacity (Metric) | 946 mL |
| Capacity (English) | 32 oz. |
| Color | Trout Green |
| Mouth | Narrow |