Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultra-high pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
- Suitable for ultra high pressure applications
- Wide chemical compatibilty
- Prefinished, square, burr-free ends and interiors to minimize dead volume connections
Specifications
Specifications
| For Use With | HPLC |
| Type | Capillary Tubing |
| Color | Black |
| For Use With (Application) | HPLC |
| Diameter (English) Inner | 0.007 in. |
| Diameter (Metric) Inner | 0.18 mm |
| Overall Length (English) | 3 ft. 3 in. |
| Overall Length (Metric) | 100 cm |
| Material | Stainless Steel |