Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC in 5-Foot Coils
Product Code. 10446754 Shop All Thermo Scientific Products
Click to view available options
Description:
0.005 in. ID; 5 ft. length
0.007 in. ID; 5 ft. length
0.010 in. ID; 5 ft. length
0.020 in. ID; 5 ft. length
0.030 in.ID; 5 ft. length
0.040 in. ID; 5 ft. length
0.046 in.ID; 5 ft. length
Unit Size:
Each
Not recommended for biological samples. Rough internal surface may lead to sample carryover. Reasonably flexible, but works best with minimal bending
Product Code. 10446754
Brand: Thermo Scientific™ U108
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Not recommended for biological samples. Rough internal surface may lead to sample carryover. Reasonably flexible, but works best with minimal bending
Use cleaned, polished and passivated Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC, ready for immediate use.
Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultrahigh-pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
  • Suitable for ultra high pressure applications
  • Wide chemical compatibilty
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC
Type Capillary Tubing
Description 0.007 in. ID; 5 ft. length
For Use With (Application) HPLC
Diameter (English) Inner 0.007 in.
Diameter (Metric) Inner 0.18 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.524 m
Material Stainless Steel