missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAMCI Antibody (OTI5E5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44948-30ul
This item is not returnable.
View return policy
Description
PAMCI Monoclonal antibody specifically detects PAMCI in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PAMCI | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 9182 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| OTI5E5 | |
| Western Blot 1:500 | |
| PAM COOH-terminal interactor protein 1, PAMCI, PCIP1, P-CIP1peptidylglycine alpha-amidating monooxygenase COOH-terminal interactorprotein-1, Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor, Ras association (RalGDS/AF-6) domain family (N-terminal) member 9, ras association domain-containing protein 9 | |
| Human recombinant protein fragment corresponding to amino acids 1-232 of human RASSF9 (NP_005438) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction