missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alas1 Antibody (OTI5D5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44908-30ul
This item is not returnable.
View return policy
Description
Alas1 Monoclonal antibody specifically detects Alas1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Alas1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5D5 | |
| Western Blot 1:2000 | |
| 5-aminolevulinic acid synthase 1, ALAS, ALAS3, ALASH, ALAS-H, aminolevulinate, delta-, synthase 1, Delta-ALA synthase 1,5-aminolevulinate synthase, nonspecific, mitochondrial, Delta-aminolevulinate synthase 1, EC 2.3.1, EC 2.3.1.37, MIG4, migration-inducing protein 4 | |
| Human recombinant protein fragment corresponding to amino acids 57-308 of human ALAS1(NP_000679) produced in E.coli. | |
| 30 μL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 211 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction