missing translation for 'onlineSavingsMsg'
Learn More

Ethacrynic Acid Epoxide, TRC

Product Code. 30370858
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370858 250 mg
30370754 1 g
30387974 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370858 Supplier Toronto Research Chemicals Supplier No. TRCE676005250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27223-10-5
Molecular Formula C13H12Cl2O5
Molecular Weight (g/mol) 319.14
Synonym [2,3-Dichloro-4-(2-ethylglycidoyl)phenoxy]acetic Acid, 2-[2,3-Dichloro-4-[(2-ethyl-2-oxiranyl)carbonyl]phenoxy]acetic Acid
SMILES ClC1=C(Cl)C(C(C2(CO2)CC)=O)=CC=C1OCC(O)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H12Cl2O5/c1-2-13(6-20-13)12(18)7-3-4-8(11(15)10(7)14)19-5-9(16)17/h3-4H,2,5-6H2,1H3,(H,16,17)
Formula Weight 318.0062
Chemical Name or Material Ethacrynic Acid Epoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.