CAS RN 27223-10-5
CAS RN 27223-10-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethacrynic Acid Epoxide, TRC
CAS: 27223-10-5 Molecular Formula: C13H12Cl2O5 Molecular Weight (g/mol): 319.14 Synonym: [2,3-Dichloro-4-(2-ethylglycidoyl)phenoxy]acetic Acid,2-[2,3-Dichloro-4-[(2-ethyl-2-oxiranyl)carbonyl]phenoxy]acetic Acid SMILES: ClC1=C(Cl)C(C(C2(CO2)CC)=O)=CC=C1OCC(O)=O