missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethylthio)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15443507
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443507 250 mg 250mg
11312499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443507 Supplier Thermo Scientific Alfa Aesar Supplier No. H26622.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 239080-04-7
Molecular Formula C9H7F3O2S
Molecular Weight (g/mol) 236.21
MDL Number MFCD00236350
InChI Key FSTITWGTGMPLTF-UHFFFAOYSA-N
Synonym 3-trifluoromethylthio phenylacetic acid, 2-3-trifluoromethyl thio phenyl acetic acid, 2-3-trifluoromethylthio phenyl acetic acid, 3-trifluoromethyl sulfanyl phenyl acetic acid, pubchem7367, acmc-1ce2m, 2-3-trifluoromethylsulfanyl phenyl acetic acid, 3-trifluoromethyl thio phenylacetic acid, benzeneacetic acid,3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl phenyl acetic acid
PubChem CID 2777897
SMILES OC(=O)CC1=CC=CC(SC(F)(F)F)=C1

Specifications

Quantity 250 mg
IUPAC Name 2-{3-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Formula Weight 236.21
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethylthio)phenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.