accessibility menu, dialog, popup

UserName

CAS RN 239080-04-7

OHOSFFF

CAS RN 239080-04-7

IUPAC Name: 2-{3-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Synonyms: {3-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Molecular Weight (g/mol): 236.21
Molecular Formula: C9H7F3O2S
InChi Key: FSTITWGTGMPLTF-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(SC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results