missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenol, 98+%

Product Code. 11405133
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11405133

Brand: Thermo Scientific Alfa Aesar A12784.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-17-9 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-17-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002299
InChI Key UGEJOEBBMPOJMT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
PubChem CID 7376
IUPAC Name 3-(trifluoromethyl)phenol
SMILES C1=CC(=CC(=C1)O)C(F)(F)F

Specifications

Melting Point -2°C
Density 1.336
Boiling Point 177°C to 178°C
Flash Point 74°C (165°F)
Odor Phenol-like
Refractive Index 1.458
Quantity 100 g
UN Number UN3265
Beilstein 2045663
Formula Weight 162.11
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)phenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.