missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)cinnamic acid, 96%, Thermo Scientific™

Product Code. p-7025856
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437154 0.25 g 0.25g
11417154 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437154 Supplier Thermo Scientific Alfa Aesar Supplier No. B21560.01

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168833-80-5
Molecular Formula C10H7F3O3
Molecular Weight (g/mol) 232.158
MDL Number MFCD00066337
InChI Key CLKZZEYGXRWYNI-SNAWJCMRSA-N
Synonym 3-trifluoromethoxy cinnamic acid, 3-3-trifluoromethoxy phenyl acrylic acid, trans-3-trifluoromethoxy cinnamic acid, 2e-3-3-trifluoromethoxy phenyl prop-2-enoic acid, rarechem bk hw 0216, 3-3-trifluoromethoxy-phenyl-acrylic acid, e-3-3-trifluoromethoxy phenyl acrylic acid, e-3-3-trifluoromethoxy phenyl prop-2-enoic acid, 3-3-trifluoromethoxy phenyl prop-2-enoic acid, pubchem4353
PubChem CID 735840
IUPAC Name (E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C=CC(=O)O

Specifications

Melting Point 92°C to 95°C
Quantity 0.25 g
Formula Weight 232.16
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethoxy)cinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.