CAS RN 168833-80-5
CAS RN 168833-80-5
IUPAC Name:
(2E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate
Synonyms:
(2E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate
Molecular Weight (g/mol):
231.15
Molecular Formula:
C10H6F3O3
InChi Key:
CLKZZEYGXRWYNI-SNAWJCMRSA-M
SMILES:
[O-]C(=O)\C=C\C1=CC=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
3-(Trifluoromethoxy)cinnamic acid, 96%, Thermo Scientific™
CAS: 168833-80-5 Molecular Formula: C10H7F3O3 Molecular Weight (g/mol): 232.158 MDL Number: MFCD00066337 InChI Key: CLKZZEYGXRWYNI-SNAWJCMRSA-N Synonym: 3-trifluoromethoxy cinnamic acid,3-3-trifluoromethoxy phenyl acrylic acid,trans-3-trifluoromethoxy cinnamic acid,2e-3-3-trifluoromethoxy phenyl prop-2-enoic acid,rarechem bk hw 0216,3-3-trifluoromethoxy-phenyl-acrylic acid,e-3-3-trifluoromethoxy phenyl acrylic acid,e-3-3-trifluoromethoxy phenyl prop-2-enoic acid,3-3-trifluoromethoxy phenyl prop-2-enoic acid,pubchem4353 PubChem CID: 735840 IUPAC Name: (E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoic acid SMILES: C1=CC(=CC(=C1)OC(F)(F)F)C=CC(=O)O