Learn More
Eppendorf™ Research 3 Neo Single Channel Pipettes Ejector Sleeve
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
2.5 μL
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
2 mL
5 mL
10 mL
Unit Size:
Each
Description
- Ejector sleeves allow for easy and efficient tip ejection
- These original Eppendorf spare parts are designed to replace the ejector sleeves of Eppendorf Research™ 3 neo pipettes
- Available in various color code with ejection carrier
Specifications
Specifications
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Ejector Sleeve |
| Volume (Metric) | 300 μL |
| For Use With (Equipment) | Pipette Tips |
| Quantity | 1 Pk. |