missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-N-[(3S)-tetrahydro-2-oxo-3-furanyl]butanamide, TRC

Product Code. 30475592
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475592 250 mg 250mg
30475986 50 mg 50mg
30472211 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475592 Supplier Toronto Research Chemicals Supplier No. TRCM282980250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1366371-61-0
Molecular Formula C9 H15 N O3
Molecular Weight (g/mol) 185.22
Synonym (S)-3-Methyl-N-(2-Oxotetrahydrofuran-3-Yl)Butanamide
IUPAC Name 3-methyl-N-[(3S)-2-oxooxolan-3-yl]butanamide
SMILES CC(C)CC(=O)N[C@H]1CCOC1=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H15NO3/c1-6(2)5-8(11)10-7-3-4-13-9(7)12/h6-7H,3-5H2,1-2H3,(H,10,11)/t7-/m0/s1
Formula Weight 185.105
Percent Purity >95%
Chemical Name or Material 3-Methyl-N-[(3S)-Tetrahydro-2-Oxo-3-Furanyl]Butanamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.