CAS RN 1366371-61-0
CAS RN 1366371-61-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methyl-N-[(3S)-tetrahydro-2-oxo-3-furanyl]butanamide, TRC
CAS: 1366371-61-0 Molecular Formula: C9 H15 N O3 Molecular Weight (g/mol): 185.22 Synonym: (S)-3-Methyl-N-(2-Oxotetrahydrofuran-3-Yl)Butanamide IUPAC Name: 3-methyl-N-[(3S)-2-oxooxolan-3-yl]butanamide SMILES: CC(C)CC(=O)N[C@H]1CCOC1=O