missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrophenol 98%, ACROS Organics™

Product Code. 10440083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10440083 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10440083 Supplier Thermo Scientific Acros Supplier No. 127301000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2581-34-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 126°C to 130°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
MDL Number MFCD00007333
Quantity 100 g
Beilstein 06, 386
Synonym 4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
InChI Key PIIZYNQECPTVEO-UHFFFAOYSA-N
SMILES CC1=CC(O)=CC=C1[N+]([O-])=O
IUPAC Name 3-methyl-4-nitrophenol
Molecular Weight (g/mol) 153.14
PubChem CID 17412
ChEBI CHEBI:38683
Formula Weight 153.14
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-nitrophenol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.