missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptoindole, TRC

Product Code. 30473025
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473025 500 mg 500mg
30472624 100 mg 100mg
30471012 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473025 Supplier Toronto Research Chemicals Supplier No. TRCM253300500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-94-4
Molecular Formula C8 H7 N S
Molecular Weight (g/mol) 149.21
IUPAC Name 1H-indole-3-thiol
SMILES Sc1c[nH]c2ccccc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H7NS/c10-8-5-9-7-4-2-1-3-6(7)8/h1-5,9-10H
Formula Weight 149.0299
Chemical Name or Material 3-Mercaptoindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.