CAS RN 480-94-4
CAS RN 480-94-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Mercaptoindole, TRC
CAS: 480-94-4 Molecular Formula: C8 H7 N S Molecular Weight (g/mol): 149.21 IUPAC Name: 1H-indole-3-thiol SMILES: Sc1c[nH]c2ccccc12