Learn More
3-Hydroxy Donepezil (Donepezil Impurity), TRC
Product Code. 30137451
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137451 100 mg 100mg
17993965 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137451 Supplier Toronto Research Chemicals Supplier No. TRCH941745100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2097683-67-3
Molecular Formula C24 H29 N O4
Molecular Weight (g/mol) 395.49
Synonym 1H-Inden-1-one, 2,3-dihydro-3-hydroxy-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, 3-Hydroxydonepezil, 2-[(1-Benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxyindan-1-one
IUPAC Name 2-[(1-benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxy-2,3-dihydroinden-1-one
SMILES COc1cc2C(O)C(CC3CCN(Cc4ccccc4)CC3)C(=O)c2cc1OC
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C24H29NO4/c1-28-21-13-18-19(14-22(21)29-2)24(27)20(23(18)26)12-16-8-10-25(11-9-16)15-17-6-4-3-5-7-17/h3-7,13-14,16,20,23,26H,8-12,15H2,1-2H3
Formula Weight 395.2097
Percent Purity >95%
Chemical Name or Material 3-Hydroxy Donepezil
Grade HPLC