CAS RN 2097683-67-3
CAS RN 2097683-67-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Hydroxy Donepezil (Donepezil Impurity), TRC
CAS: 2097683-67-3 Molecular Formula: C24 H29 N O4 Molecular Weight (g/mol): 395.49 Synonym: 1H-Inden-1-one, 2,3-dihydro-3-hydroxy-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-,3-Hydroxydonepezil,2-[(1-Benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxyindan-1-one IUPAC Name: 2-[(1-benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxy-2,3-dihydroinden-1-one SMILES: COc1cc2C(O)C(CC3CCN(Cc4ccccc4)CC3)C(=O)c2cc1OC
2
2-[(1-Benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxyindan-1-one (3-Hydroxydonepezil), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.