Learn More
3-Hydroxy-1-methylpyridinium Bromide, Mikromol™
Product Code. 30050151
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050151 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30050151 Supplier Mikromol Supplier No. MM0429.040025
 Restricted Item
Estimated Shipment: 30-10-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Hydroxy-1-methylpyridinium Bromide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hydroxy-1-methylpyridin-1-ium bromide
CAS 31034-86-3
Analyte or Component Names 3-Hydroxy-1-methylpyridin-1-ium bromide
Shipping Condition Ice pack (-20°C)
Molecular Formula C6H8BrNO
Certifications/Compliance ISO 9001
Synonym Pyridostigmine Related Compound B, 3-Hydroxy-1-methylpyridinium Bromide, Pyridinium, 3-hydroxy-1-methyl-, bromide (1:1), 3-Hydroxy-1-methylpyridinium bromide (6CI), Pyridinium, 3-hydroxy-1-methyl-, bromide (8CI,9CI), 3-Hydroxy-N-methylpyridinium bromide, N-Methyl-3-hydroxypyridinium bromide, Ro 1-5237
InChI Formula InChI=1S/C6H7NO.BrH/c1-7-4-2-3-6(8)5-7;/h2-5H,1H3;1H
SMILES C[N+]1=CC=CC(=C1)O.[Br-]
IUPAC Name 1-methylpyridin-1-ium-3-ol;bromide
Molecular Weight (g/mol) 190.04
Quantity 25 mg
Formula Weight 188.979 g/mol