Learn More
3-Fluoro-5-hydroxybenzeneboronic acid, 98%, Thermo Scientific™
Product Code. p-7034918
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15449377
Brand: Thermo Scientific Alfa Aesar H52619.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Fluoro-5-hydroxybenzeneboronic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 871329-82-7
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.919
MDL Number MFCD07363778
InChI Key RMBFBZIEXCTPDB-UHFFFAOYSA-N
Synonym 3-fluoro-5-hydroxyphenyl boronic acid, 3-fluoro-5-hydroxybenzeneboronic acid, 3-fluoro-5-hydroxy-phenyl boronic acid, 3-fluoro-5-hydroxphenyl boronic acid, 3-hydroxyl-5-fluoro-phenyl-boronic acid, acmc-209qgp, 3-fluoro-5-hydroxyphenyl-boronic acid, 3-fluoranyl-5-oxidanyl-phenyl boronic acid, boronic acid,b-3-fluoro-5-hydroxyphenyl
PubChem CID 44717308
IUPAC Name (3-fluoro-5-hydroxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)O)(O)O
Specifications
Melting Point 54°C to 62°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 155.92
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-hydroxybenzeneboronic acid