Learn More
3-Fluoro-4-(trifluoromethyl)aniline, 97%
Product Code. 11415867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415867 5 g 5g
11405867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11415867 Supplier Thermo Scientific Alfa Aesar Supplier No. B24411.06
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
CAS: 69411-68-3 | C7H5F4N | 179.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69411-68-3
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00190120
InChI Key CRRVZRDISHOQQL-UHFFFAOYSA-N
Synonym 4-amino-2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl aniline, 2-fluoro-4-amino benzotrifluoride, benzenamine, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl-phenylamine, 3-fluoro-4-trifluoromethyl phenylamine, pubchem1631, 4-amino,2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl-aniline
PubChem CID 2735914
IUPAC Name 3-fluoro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C(F)=C1)C(F)(F)F
Specifications
Melting Point 54°C to 58°C
Quantity 5 g
UN Number UN2811
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)aniline