missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11429887
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429887 1 g 1g
11439887 5 g 5g
11369489 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429887 Supplier Thermo Scientific Alfa Aesar Supplier No. B22846.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59189-97-8
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD00671774
InChI Key DPWCZSXNEGNALT-UHFFFAOYSA-N
PubChem CID 2734875
IUPAC Name 3-fluoro-4-methylbenzoyl chloride
SMILES CC1=CC=C(C=C1F)C(Cl)=O

Specifications

Density 1.25
Refractive Index 1.5325
Quantity 1 g
UN Number UN3265
Beilstein 7757348
Sensitivity Moisture sensitive
Synonym 3-fluoro-4-methyl-benzoyl chloride, 3-fluoro-4-methylbenzoylchloride, acmc-20an3p, 3-fluoro-4-methyl-benzoylchloride, 3-fluoro-4-methyl benzoyl chloride, benzoyl chloride,3-fluoro-4-methyl, 3-fluoranyl-4-methyl-benzoyl chloride, benzoyl chloride, 3-fluoro-4-methyl, 3-fluoro-4-methylbenzoyl chloride, benzoyl chloride, 3-fluoro-4-methyl-9ci
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.