accessibility menu, dialog, popup

UserName

CAS RN 59189-97-8

CH3FClO

CAS RN 59189-97-8

IUPAC Name: 3-fluoro-4-methylbenzoyl chloride
Synonyms: 3-fluoro-4-methylbenzoyl chloride
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: DPWCZSXNEGNALT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1F)C(Cl)=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results