Learn More
3-Ethylpyridine, 98%
Product Code. 11439836
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11439836
Brand: Thermo Scientific Alfa Aesar B20318.06
Request Bulk or Custom Format
CAS: 536-78-7 | C7H9N | 107.156 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 536-78-7
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006413
InChI Key MFEIKQPHQINPRI-UHFFFAOYSA-N
Synonym pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394
PubChem CID 10823
IUPAC Name 3-ethylpyridine
SMILES CCC1=CN=CC=C1
Specifications
Melting Point -77°C
Boiling Point 165°C to 166°C
Flash Point 49°C (120°F)
Odor Unpleasant
Quantity 5 g
UN Number UN1993
Beilstein 106479
Merck Index 14,3848
Formula Weight 107.16
Percent Purity 98%
Chemical Name or Material 3-Ethylpyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Ethylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-Ethylpyridine