missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3’-Desmethylamino-3’-(methanesulfonyl)hydroxy-10-hydroxy Dehydroxy-hydro-nortriptyline, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | 3’-Desmethylamino-3’-(methanesulfonyl)hydroxy-10-hydroxy Dehydroxy-hydro-nortriptyline |
| Analyte or Component Names | 3’-Desmethylamino-3’-(methanesulfonyl)hydroxy-10-hydroxy Dehydroxy-hydro-nortriptyline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19H18O3S |
| Synonym | 3-(5H-Dibenzo[a,d][7]annulen-5-ylidene)propyl Methanesulfonate |
| InChI Formula | InChI=1S/C19H18O3S/c1-23(20,21)22-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19/h2-5,7-13H,6,14H2,1H3 |
| Solution Type | Neat |
| SMILES | CS(=O)(OCC/C=C1C2=CC=CC=C2C=CC3=CC=CC=C/13)=O |
| IUPAC Name | 3-(5H-dibenzo[a,d][7]annulen-5-ylidene)propyl methanesulfonate |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction