missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroadamantane-1-carboxylic acid, 97%

Product Code. 11448917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448917 1 g 1g
11458917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448917 Supplier Thermo Scientific Alfa Aesar Supplier No. B20543.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34859-74-0 | C11H15ClO2 | 214.689 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34859-74-0
Molecular Formula C11H15ClO2
Molecular Weight (g/mol) 214.689
MDL Number MFCD00167831
InChI Key GMOBHRQSNDDZAD-UHFFFAOYSA-N
Synonym 1-chloro-3-adamantanecarboxylic acid, 3-chloroadamantanecarboxylic acid, 3-adamantanecarboxylic acid, 1-chloro, 3-chlorotricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, acmc-1advo, maybridge1_005634, 1r,3s,5r,7s-3-chloroadamantane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3-chloro, n-4-3-4-methoxyphenyl propanoyl-2,3,4,5-tetrahydro-1,4-benzoxazepin-7-yl methyl-n-phenylcyclopropanecarboxamide
PubChem CID 36971
IUPAC Name 3-chloroadamantane-1-carboxylic acid
SMILES C1C2CC3(CC1CC(C2)(C3)Cl)C(=O)O

Specifications

Quantity 1 g
Formula Weight 214.69
Percent Purity 97%
Chemical Name or Material 3-Chloroadamantane-1-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.