accessibility menu, dialog, popup

UserName

CAS RN 34859-74-0

IUPAC Name: (5R,7S)-3-chloroadamantane-1-carboxylate
Synonyms: (5R,7S)-3-chloroadamantane-1-carboxylate
Molecular Weight (g/mol): 213.68
Molecular Formula: C11H14ClO2
InChi Key: GMOBHRQSNDDZAD-JZVMUCMXSA-M
SMILES: [O-]C(=O)C12C[C@H]3C[C@H](CC(Cl)(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results