missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-methylanisole, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Tamaño de la unidad:
100g
25g
5g
Descripción
Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Especificaciones
Especificaciones
| Melting Point | -3°C |
| Density | 1.16 |
| Boiling Point | 212°C to 215°C |
| Flash Point | 92°C (197°F) |
| Refractive Index | 1.5395 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Beilstein | 1862220 |
| Formula Weight | 156.61 |
| Percent Purity | 97% |
| Mostrar más |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido