accessibility menu, dialog, popup

UserName

H3COClH3C

CAS RN 3260-88-6

IUPAC Name: 1-chloro-3-methoxy-2-methylbenzene
Synonyms: 1-chloro-3-methoxy-2-methylbenzene
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: LTVRGAWOEOKGJZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(Cl)=C1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results