Learn More
3'-Bromopropiophenone, 97%
Product Code. 11469230
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11469230
Brand: Thermo Scientific Alfa Aesar A16757.06
Request Bulk or Custom Format
CAS: 19829-31-3 | C9H9BrO | 213.074 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19829-31-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000084
InChI Key QSHLXVTVXQTHBS-UHFFFAOYSA-N
Synonym 3'-bromopropiophenone, 1-3-bromophenyl propan-1-one, 1-propanone, 1-3-bromophenyl, m-bromopropiophenone, 1-3-bromophenyl-1-propanone, 3-bromophenyl ethyl ketone, attercop-chm at106303, pubchem19020, acmc-209f2u
PubChem CID 88272
IUPAC Name 1-(3-bromophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Br
Specifications
Melting Point 38°C to 41°C
Boiling Point 144°C to 145°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1936736
Formula Weight 213.08
Percent Purity 97%
Chemical Name or Material 3'-Bromopropiophenone