accessibility menu, dialog, popup

UserName

CAS RN 19829-31-3

CH3OBr

CAS RN 19829-31-3

IUPAC Name: 1-(3-bromophenyl)propan-1-one
Synonyms: 1-(3-bromophenyl)propan-1-one
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: QSHLXVTVXQTHBS-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)