missing translation for 'onlineSavingsMsg'
Learn More

3-[5-Fluoro-2-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Product Code. 15468095
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468095 250 mg 250mg
15458095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468095 Supplier Thermo Scientific Alfa Aesar Supplier No. H32457.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-43-4
Molecular Formula C10H8F4O2
Molecular Weight (g/mol) 236.166
MDL Number MFCD09025360
InChI Key JHDJFXXGNKFTSB-UHFFFAOYSA-N
Synonym 3-5-fluoro-2-trifluoromethyl phenyl propionic acid, 3-5-fluoro-2-trifluoromethyl phenyl propanoic acid
PubChem CID 46737491
IUPAC Name 3-[5-fluoro-2-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1F)CCC(=O)O)C(F)(F)F

Specifications

Melting Point 63°C to 66°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 236.16
Percent Purity 97%
Chemical Name or Material 3-[5-Fluoro-2-(trifluoromethyl)phenyl]propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.