accessibility menu, dialog, popup

UserName

CAS RN 916420-43-4

FOHOFFF

CAS RN 916420-43-4

IUPAC Name: 3-[5-fluoro-2-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[5-fluoro-2-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 236.17
Molecular Formula: C10H8F4O2
InChi Key: JHDJFXXGNKFTSB-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C(C=CC(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results