missing translation for 'onlineSavingsMsg'
Learn More

3-[5-Fluoro-2-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Código de producto. 15458095
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
15458095 1 g 1g
15468095 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15458095 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H32457.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids, bases.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 916420-43-4
Molecular Formula C10H8F4O2
Molecular Weight (g/mol) 236.166
MDL Number MFCD09025360
InChI Key JHDJFXXGNKFTSB-UHFFFAOYSA-N
Synonym 3-5-fluoro-2-trifluoromethyl phenyl propionic acid, 3-5-fluoro-2-trifluoromethyl phenyl propanoic acid
PubChem CID 46737491
IUPAC Name 3-[5-fluoro-2-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1F)CCC(=O)O)C(F)(F)F

Especificaciones

Melting Point 63°C to 66°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 236.16
Percent Purity 97%
Chemical Name or Material 3-[5-Fluoro-2-(trifluoromethyl)phenyl]propionic acid
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.