missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxy-4-methylbenzoic acid, 97%

Product Code. 11315259
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11315259

Brand: Thermo Scientific Alfa Aesar A19196.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28026-96-2 | C8H7O4 | 167.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28026-96-2
Molecular Formula C8H7O4
Molecular Weight (g/mol) 167.14
MDL Number MFCD00002514
InChI Key KMRRXSZDSGYLCD-UHFFFAOYSA-M
PubChem CID 2748045
IUPAC Name 3,5-dihydroxy-4-methylbenzoic acid
SMILES CC1=C(O)C=C(C=C1O)C([O-])=O

Specifications

Melting Point 266°C to 207°C
Quantity 5 g
Synonym 3,5-dihydroxy-p-toluic acid, benzoic acid, 3,5-dihydroxy-4-methyl, 4-methyl-3,5-dihydroxy benzoic acid, 3,5-dihydroxy-4-methyl-benzoic acid, pubchem3868, acmc-20a50j, paragos 440034, 3,5-dihydroxy4-methylbenzoic acid, 3,5-dihydroxy-4-methyl benzoic acid, benzoic acid,3,5-dihydroxy-4-methyl
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,5-Dihydroxy-4-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.