Learn More
3,5-Bis(trifluoromethyl)benzhydrazide, 97%
Product Code. 11465583
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11465583 25 g 25g
11445583 1 g 1g
11455583 5 g 5g
3 options
Product Code. 11465583
Brand: Thermo Scientific Alfa Aesar A13492.14
Request Bulk or Custom Format
CAS: 26107-82-4 | C9H6F6N2O | 272.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26107-82-4
Molecular Formula C9H6F6N2O
Molecular Weight (g/mol) 272.15
MDL Number MFCD00051848
InChI Key GBBRFBSFWKFTMZ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzhydrazide, 3,5-bis trifluoromethyl benzohydrazide, 3,5-di trifluoromethyl benzhydrazide, 3,5-bis-trifluoromethylbenzoic acid hydrazide, 3,5-bis trifluoromethyl benzoic acid hydrazide, benzoic acid, 3,5-bis trifluoromethyl-, hydrazide, 3,5-bis-trifluoromethyl-benzhydrazide, maybridge1_001057, acmc-1cp6z, ksc206c6d
PubChem CID 100265
IUPAC Name 3,5-bis(trifluoromethyl)benzohydrazide
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)NN
Specifications
Melting Point 132°C to 134°C
Quantity 25 g
Beilstein 2596448
Formula Weight 272.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzhydrazide