missing translation for 'onlineSavingsMsg'
Learn More

3,5,6-Trifluoro-2-hydroxypyridine, 98%, Thermo Scientific™

Product Code. 15465867
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15465867

Brand: Thermo Scientific Alfa Aesar H33085.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75777-49-0
Molecular Formula C5H2F3NO
Molecular Weight (g/mol) 149.072
MDL Number MFCD04114299
InChI Key PWBGZZXJAMGVGQ-UHFFFAOYSA-N
Synonym 3,5,6-trifluoro-2-hydroxypyridine, 2-hydroxy-3,5,6-trifluoropyridine, 3,5,6-trifluoropyridin-2-ol, acmc-1beik, 3,5,6-trifluoro-2-pyridone, 3,5,6-trifluoro-2-pyridinol, 3,5,6-trifluoropyridin-2 1h-one, 2 1h-pyridinone,3,5,6-trifluoro, 3,5,6-trifluoro-1,2-dihydropyridine-2-one, 2 1h-pyridinone, 3,5,6-trifluoro-9ci
PubChem CID 23340297
IUPAC Name 3,5,6-trifluoro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC(=C1F)F)F

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
Formula Weight 149.07
Percent Purity 98%
Chemical Name or Material 3,5,6-Trifluoro-2-hydroxypyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.