accessibility menu, dialog, popup

UserName

CAS RN 75777-49-0

OHNFFF

CAS RN 75777-49-0

IUPAC Name: 3,5,6-trifluoro-1,2-dihydropyridin-2-one
Synonyms: 3,5,6-trifluoro-1H-pyridin-2-one
Molecular Weight (g/mol): 149.07
Molecular Formula: C5H2F3NO
InChi Key: PWBGZZXJAMGVGQ-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(F)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results