Learn More
3,4-Dimethyl-1-(1-chloroethyl)benzene (>85%), TRC
Product Code. 30180424
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180424 100 mg 100mg
30179950 50 mg 50mg
30180362 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30180424 Supplier Toronto Research Chemicals Supplier No. TRCD471833100MG
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104245-83-2
Molecular Formula C10 H13 Cl
Molecular Weight (g/mol) 168.66
Synonym Medetomidine Impurity 30
IUPAC Name 4-(1-chloroethyl)-1,2-dimethylbenzene
SMILES ClC(C)C1=CC=C(C)C(C)=C1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H13Cl/c1-7-4-5-10(9(3)11)6-8(7)2/h4-6,9H,1-3H3
Formula Weight 168.07
Chemical Name or Material 3,4-Dimethyl-1-(1-chloroethyl)benzene