CAS RN 104245-83-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dimethyl-1-(1-chloroethyl)benzene (>85%), TRC
CAS: 104245-83-2 Molecular Formula: C10 H13 Cl Molecular Weight (g/mol): 168.66 Synonym: Medetomidine Impurity 30 IUPAC Name: 4-(1-chloroethyl)-1,2-dimethylbenzene SMILES: ClC(C)C1=CC=C(C)C(C)=C1