Learn More
3',4'-Dihydrospiro[androst-4,6-diene-17,2'(5'H)-furan]-3,5'-dione (Canrenone), Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
3',4'-Dihydrospiro[androst-4,6-diene-17,2'(5'H)-furan]-3,5'-dione (Canrenone)
Specifications
Specifications
| Chemical Name or Material | Canrenone |
| CAS | 976-71-6 |
| Analyte or Component Names | Canrenone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Spironolactone Imp. F (EP), Spironolactone USP Related Compound A (AS), Spironolactone USP RC A (AS), (2'R)-3',4'-Dihydro-5'H-spiro[androst-4,6-diene-17,2'-furan]-3,5'-dione, Pregna-4,6-diene-21-carboxylic acid, 17-hydroxy-3-oxo-, gamma-lactone, (17alpha)-, 17alpha-Pregna-4,6-diene-21-carboxylic acid, 17-hydroxy-3-oxo-, gamma-lactone (6CI,7CI,8CI), Spiro[17H-cyclopenta[a]phenanthrene-17,2'(5'H)-furan], pregna-4,6-diene-21-carboxylic acid deriv., 11614 R.P., 17-Hydroxy-3-oxo-17alpha-pregna-4,6-diene-21-carboxylic acid lactone, 17-Hydroxy-3-oxo-17alpha-pregna-4,6-diene-21-carboxylic acid gamma-lactone, 17alpha-(2-Carboxyethyl)-17beta-hydroxyandrosta-4,6-dien-3-one lactone, 17beta-Hydroxy-3-oxopregna-4,6-diene-21-carboxylic acid, 20-Spiroxa-4,6-diene-3,21-dione, 3-(17beta-Hydroxy-3-oxoandrosta-4,6-dien-17alpha-yl)propionic acid lactone, 3-(17beta-Hydroxy-3-oxoandrosta-4,6-dien-17alpha-yl)propionic acid gamma-lactone, 3-(3-Oxo-17beta-hydroxy-4,6-androstadien-17alpha-yl)propionic acid gamma-lactone, 3'-(3-Oxo-17beta-hydroxyandrosta-4,6-dien-17alpha-yl)-propionic acid lactone, Aldadiene, Canrenone, NSC 261713, Phanurane, SC 9376, Spirolactone SC 14266, Spironolactone Related Compound A, Spironolactone Impurity F |
| InChI Formula | InChI=1S/C22H28O3/c1-20-9-5-15(23)13-14(20)3-4-16-17(20)6-10-21(2)18(16)7-11-22(21)12-8-19(24)25-22/h3-4,13,16-18H,5-12H2,1-2H3/t16-,17+,18+,20+,21+,22-/m1/s1 |
| SMILES | C[C@]12CCC(=O)C=C1C=C[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]45CCC(=O)O5 |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
| Show More |