Learn More
3,4-Difluoro-2-methylbenzonitrile, 98%, Thermo Scientific™
Product Code. 15434147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434147 5 g 5g
15424147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15434147 Supplier Thermo Scientific Alfa Aesar Supplier No. H31548.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 847502-83-4
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD07777154
InChI Key LMGHUTIUVKVZNH-UHFFFAOYSA-N
PubChem CID 24820441
IUPAC Name 3,4-difluoro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1F)F)C#N
Specifications
Melting Point 39°C to 44°C
Quantity 5 g
UN Number UN3439
Synonym benzonitrile, 3,4-difluoro-2-methyl, 3,4-difluoro-o-tolunitrile, benzonitrile,3,4-difluoro-2-methyl, 2,3-difluoro-6-cyano-toluene, 3,4-difluoro-2-methyl benzonitrile, pubchem4769, 3,4-difluoro-2-methyl-benzonitrile
Formula Weight 153.13
Percent Purity 98%
Chemical Name or Material 3,4-Difluoro-2-methylbenzonitrile