accessibility menu, dialog, popup

UserName

CAS RN 847502-83-4

CH3FFN

CAS RN 847502-83-4

IUPAC Name: 3,4-difluoro-2-methylbenzonitrile
Synonyms: 3,4-difluoro-2-methylbenzonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: LMGHUTIUVKVZNH-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(F)=C1F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results