missing translation for 'onlineSavingsMsg'
Learn More

3,4-Bis(phenylmethoxy)benzeneethanol 1-Acetate, TRC

Product Code. 30335832
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335832 50 mg 50mg
30333008 25 mg 25mg
30345857 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30335832 Supplier Toronto Research Chemicals Supplier No. TRCB52147550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Bis(phenylmethoxy)benzeneethanol 1-Acetate
Analyte or Component Names 3,4-Bis(phenylmethoxy)benzeneethanol 1-Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H24 O4
InChI Formula InChI=1S/C24H24O4/c1-19(25)26-15-14-20-12-13-23(27-17-21-8-4-2-5-9-21)24(16-20)28-18-22-10-6-3-7-11-22/h2-13,16H,14-15,17-18H2,1H3
Solution Type Neat
SMILES CC(=O)OCCc1ccc(OCc2ccccc2)c(OCc3ccccc3)c1
IUPAC Name 2-[3,4-bis(phenylmethoxy)phenyl]ethyl acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 376.445
Quantity 50 mg
Formula Weight 376.167
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.