Learn More
3,4,5,6,7,8-Hexahydrocoumarin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30324954
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324954 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324954 Supplier Dr. Ehrenstorfer Supplier No. DREA14194300AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Hexahydrocoumarin
CAS 700-82-3
Analyte or Component Names Hexahydrocoumarin
Shipping Condition +20°C
Molecular Formula C9 H12 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Coumarin, 3,4,5,6,7,8-hexahydro- (6CI,7CI,8CI), 3,4,5,6,7,8-Hexahydro-2H-1-benzopyran-2-one, 1-Cyclohexene-1-propanoic acid, 2-hydroxy-, Δ-lactone, 3,4,5,6,7,8-Hexahydrocoumarin, Hexahydrocoumarin, Δ-1,6-2-Oxabicyclo(4.4.0)decen-3-one
InChI Formula InChI=1S/C9H12O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H2
SMILES O=C1CCC2=C(CCCC2)O1
IUPAC Name 3,4,5,6,7,8-hexahydrochromen-2-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 152.19
Quantity 1 mL
Formula Weight 152.0837 g/mol