missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Trifluoromethyl)phenyl]propanol, Mikromol™

Product Code. 17960346
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17960346

Brand: Mikromol MM3370.080100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[3-(Trifluoromethyl)phenyl]propanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propanol
CAS 78573-45-2
Analyte or Component Names 3-[3-(Trifluoromethyl)phenyl]propanol
Shipping Condition Room Temperature
Molecular Formula C10 H11 F3 O
Certifications/Compliance ISO 9001
Synonym Benzenepropanol, 3-(trifluoromethyl)-, 3-(Trifluoromethyl)benzenepropanol, 3-(3'-Trifluoromethylphenyl)propan-1-ol, 3-[3-(Trifluoromethyl)phenyl]propan-1-ol, Cinacalcet Related Compound E
InChI Formula InChI=1S/C10H11F3O/c11-10(12,13)9-5-1-3-8(7-9)4-2-6-14/h1,3,5,7,14H,2,4,6H2
SMILES OCCCc1cccc(c1)C(F)(F)F
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propan-1-ol
Molecular Weight (g/mol) 204.1889
Quantity 100 mg
Formula Weight 204.0762 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.