Learn More
3,3’,5-Triiodo-L-thyronine 4’-O-Sulfate, TRC
Product Code. 30007063
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007063 2.5 mg 2.5mg
30006037 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30007063 Supplier Toronto Research Chemicals Supplier No. TRCT7953952.5MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31135-55-4
Molecular Formula C15H12I3NO7S
Molecular Weight (g/mol) 731.04
Synonym Triiodothyronine Sulfate, 3,5-Diiodo-O-[3-iodo-4-(sulfooxy)phenyl]-L-tyrosine, 3-[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-L-alanine Hydrogen Sulfate, Triiodothyronine Hydrogen Sulfate, Liothyronine Sulfate, T3 Sulfate;
IUPAC Name (2S)-2-amino-3-[3,5-diiodo-4-(3-iodo-4-sulfooxyphenoxy)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1OC2=C(C=C(C=C2I)C[C@@H](C(=O)O)N)I)I)OS(=O)(=O)O
Specifications
InChI Formula InChI=1S/C15H12I3NO7S/c16-9-6-8(1-2-13(9)26-27(22,23)24)25-14-10(17)3-7(4-11(14)18)5-12(19)15(20)21/h1-4,6,12H,5,19H2,(H,20,21)(H,22,23,24)/t12-/m0/s1
Quantity 2.5 mg
Formula Weight 730.7469
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3,3',5-Triiodo-L-thyronine 4'-O-Sulfate
Grade HPLC